missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PKIA Full-length ORF (NP_006814.1, 1 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005569-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119551

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the cAMP-dependent protein kinase (PKA) inhibitor family. This protein was demonstrated to interact with and inhibit the activities of both C alpha and C beta catalytic subunits of the PKA. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]

Sequence: MTDVETTYADFIASGRTGRRNAIHDILVSSASGNSNELALKLAGLDINKTEGEEDAQRSSTEQSGEAQGEAAKSES
Specifications

Specifications

NP_006814.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRKACN1
PKIA
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5569
PKIA (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTDVETTYADFIASGRTGRRNAIHDILVSSASGNSNELALKLAGLDINKTEGEEDAQRSSTEQSGEAQGEAAKSES
RUO
PKIA
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers