missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PIP5K2A Partial ORF (NP_005019, 304 a.a. - 365 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005305-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102855

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Phosphatidylinositol-5,4-bisphosphate, the precursor to second messengers of the phosphoinositide signal transduction pathways, is thought to be involved in the regulation of secretion, cell proliferation, differentiation, and motility. The protein encoded by this gene is one of a family of enzymes capable of catalyzing the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. The amino acid sequence of this enzyme does not show homology to other kinases, but the recombinant protein does exhibit kinase activity. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. [provided by RefSeq]

Sequence: SDGTHPVGTPPDSPGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKEVYFMAIIDILTHYD
Specifications

Specifications

NP_005019
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.56kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SDGTHPVGTPPDSPGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKEVYFMAIIDILTHYD
RUO
PIP4K2A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5305
PIP5K2A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13267/PI5P4KA/PIP5K2A/PIP5KII-alpha/PIP5KIIA/PIPK
PIP4K2A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers