Learn More
Abnova™ Human PIP5K1C Full-length ORF (AAH11138.1, 1 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023396-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the type I phosphatidylinositol-4-phosphate 5-kinase family of enzymes. A similar protein in mice is found in synapses and focal adhesion plaques, and binds the FERM domain of talin through its C-terminus. [provided by RefSeq]
Sequence: MTPRHPLFTSRRPPAQPTRCARKAPAVEKPDPSPRSPARTHTPREQPLHSAQERGRPSSGTGQPRGSQHPSGQAHVGSSLRAQRVCPALGSDLGLSSQPGAHLLAPGIHPSAPALSLRRTPSRHPPGRQGQRSEGVSGTWMASRDRGIVISKAQPPEATPVLPGSPRVALPRALAPETPRACGVTRSSRCGPGRAAPRPGQPPSAESpecifications
AAH11138.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0589/LCCS3/PIP5K-GAMMA/PIP5Kgamma | |
PIP5K1C | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23396 | |
PIP5K1C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTPRHPLFTSRRPPAQPTRCARKAPAVEKPDPSPRSPARTHTPREQPLHSAQERGRPSSGTGQPRGSQHPSGQAHVGSSLRAQRVCPALGSDLGLSSQPGAHLLAPGIHPSAPALSLRRTPSRHPPGRQGQRSEGVSGTWMASRDRGIVISKAQPPEATPVLPGSPRVALPRALAPETPRACGVTRSSRCGPGRAAPRPGQPPSAE | |
RUO | |
PIP5K1C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |