missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PIGX Full-length ORF (NP_060331.1, 1 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054965-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16170983

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Glycosylphosphatidylinositol (GPI) is a complex glycolipid that anchors many proteins to the cell surface. PIGX is a subunit of a GPI mannosyltransferase complex involved in the synthesis of the core GPI structure in the endoplasmic reticulum (ER) (Ashida et al., 2005 [PubMed 15635094]).[supplied by OMIM]

Sequence: MCSEIILRQEVLKDGFHRDLLIKVKFGESIEDLHTCRLLIKQDIPAGLYVDPYELASLRERNITEAVMVSENFDIEAPNYLSKESEVLIYARRDSQCIDCFQAFLPVHCRYHRPHSEDGEASIVVNNPDLLMFCDQEFPILKCWAHSEVAAPCALDNEDICQWNKMKYKSVYKNVILQVPVGLTVHTSLVCSVTLLITILCSTLILVAVFKYGHFSL
Specifications

Specifications

NP_060331.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20522
PIGX
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54965
PIGX (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MCSEIILRQEVLKDGFHRDLLIKVKFGESIEDLHTCRLLIKQDIPAGLYVDPYELASLRERNITEAVMVSENFDIEAPNYLSKESEVLIYARRDSQCIDCFQAFLPVHCRYHRPHSEDGEASIVVNNPDLLMFCDQEFPILKCWAHSEVAAPCALDNEDICQWNKMKYKSVYKNVILQVPVGLTVHTSLVCSVTLLITILCSTLILVAVFKYGHFSL
RUO
PIGX
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers