Learn More
R&D Systems™ Human PIAS3 (NP_006090.2) VersaClone cDNA
Product Code. 18752147
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752147 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18752147 Supplier R&D Systems™ Supplier No. RDC3766
  • ONLINE EXCLUSIVE PRICE
    853.00€
    806.40€ / 10µg
    Save 46.60€
    5% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Specifikationer
Gene PIAS3
Accession Number NP_006090.2
Gene ID (Entrez) 10401
Gene Alias MIZ-type containing 5, Protein inhibitor of activated STAT protein 3, protein inhibitor of activated STAT, 3, ZMIZ5
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.