missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PHLDA3 Full-length ORF (NP_036528.1, 1 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023612-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127542

  • 359.00€ / 10µg

Estimated Shipment: 06-10-2022
to see stock.
Add to basket

Description

Description

Sequence: MTAAATATVLKEGVLEKRSGGLLQLWKRKRCVLTERGLQLFEAKGTGGRPKELSFARIKAVECVESTGRHIYFTLVTEGGGEIDFRCPLEDPGWNAQITLGLVKFKNQQAIQTVRARQSLGTGTLVS
Specifications

Specifications

NP_036528.1
Liquid
23612
PHLDA3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAAATATVLKEGVLEKRSGGLLQLWKRKRCVLTERGLQLFEAKGTGGRPKELSFARIKAVECVESTGRHIYFTLVTEGGGEIDFRCPLEDPGWNAQITLGLVKFKNQQAIQTVRARQSLGTGTLVS
RUO
PHLDA3
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIH1
PHLDA3
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers