Learn More
Abnova™ Human PHKA1 Partial ORF (NP_002628, 631 a.a. - 730 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005255-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The PHKA1 gene encodes the alpha subunit of muscle phosphorylase kinase (EC 2.7.1.38), a key regulatory enzyme of glycogen metabolism. Phosphorylase kinase consists of 4 copies of an alpha-beta-gamma-delta tetramer. The alpha, beta (PHKB; MIM 172490), and gamma (PHKG1; MIM 172470 and PHKG2; MIM 172471) subunits have several isoforms; the delta subunit is calmodulin (CALM1; MIM 114180). PHKA2 (MIM 306000) encodes the alpha subunit of liver-specific phosphorylase kinase and is also located on the X chromosome.[supplied by OMIM]
Sequence: DYDDNYDYLESGNWMNDYDSTSHARCGDEVARYLDHLLAHTAPHPKLAPTSQKGGLDRFQAAVQTTCDLMSLVTKAKELHVQNVHMYLPTKLFQASRPSFSpecifications
NP_002628 | |
Liquid | |
5255 | |
PHKA1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC132604/PHKA | |
PHKA1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DYDDNYDYLESGNWMNDYDSTSHARCGDEVARYLDHLLAHTAPHPKLAPTSQKGGLDRFQAAVQTTCDLMSLVTKAKELHVQNVHMYLPTKLFQASRPSF | |
RUO | |
PHKA1 | |
Wheat Germ (in vitro) | |
GST |