missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PHF15 Full-length ORF (AAH04292.1, 1 a.a. - 246 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

349.00€ - 528.00€

Specifications

Accession Number AAH04292.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23338
Molecular Weight (g/mol) 52.3kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16107182
View Documents Promotion Details
Abnova™
H00023338-P01.25UG
25 ug
528.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
16197172
View Documents Promotion Details
Abnova™
H00023338-P01.10UG
10 ug
349.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
Description

Description

Sequence: MAMSEWPLNNGHREDPAPGLLSEELLQDEETLLSFMRDPSLRPGDPARKARGRTRLPAKKKPPPPPPQDGPGSRTTPDKAPKKTWGQDAGSGKGGQGPPTRKPPRRTSSHLPSSPAAGDCPILATPESPPPLAPETPDEAASVAADSDVQVPGPAASPKPLGRLRPPRESKVTRRLPGARPDAGMGPPSAVAERPKISLHFDTETDGYFSDGEMSDSDVEAEDGGVQRGPREAGAEEVVRMGVLAS
Specifications

Specifications

AAH04292.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.3kDa
Glutathione Sepharose 4 Fast Flow
MAMSEWPLNNGHREDPAPGLLSEELLQDEETLLSFMRDPSLRPGDPARKARGRTRLPAKKKPPPPPPQDGPGSRTTPDKAPKKTWGQDAGSGKGGQGPPTRKPPRRTSSHLPSSPAAGDCPILATPESPPPLAPETPDEAASVAADSDVQVPGPAASPKPLGRLRPPRESKVTRRLPGARPDAGMGPPSAVAERPKISLHFDTETDGYFSDGEMSDSDVEAEDGGVQRGPREAGAEEVVRMGVLAS
RUO
PHF15
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
23338
PHF15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
JADE2/KIAA0239
PHF15
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications