Learn More
Abnova™ Human PGAM2 Full-length ORF (NP_000281.2, 1 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005224-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Phosphoglycerate mutase (PGAM) catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). This gene encodes muscle-specific PGAM subunit. Mutations in this gene cause muscle phosphoglycerate mutase eficiency, also known as glycogen storage disease X. [provided by RefSeq]
Sequence: MATHRLVMVRHGESTWNQENRFCGWFDAELSEKGTEEAKRGAKAIKDAKMEFDICYTSVLKRAIRTLWAILDGTDQMWLPVVRTWRLNERHYGGLTGLNKAETAAKHGEEQVKIWRRSFDIPPPPMDEKHPYYNSISKERRYAGLKPGELPTCESLKDTIARALPFWNEEIVPQIKAGKRVLIAAHGNSLRGIVKHLEGMSDQAIMELNLPTGIPIVYELNKELKPTKPMQFLGDEETVRKAMEAVAAQGKAKSpecifications
NP_000281.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC88743/PGAM-M/PGAMM | |
PGAM2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5224 | |
PGAM2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATHRLVMVRHGESTWNQENRFCGWFDAELSEKGTEEAKRGAKAIKDAKMEFDICYTSVLKRAIRTLWAILDGTDQMWLPVVRTWRLNERHYGGLTGLNKAETAAKHGEEQVKIWRRSFDIPPPPMDEKHPYYNSISKERRYAGLKPGELPTCESLKDTIARALPFWNEEIVPQIKAGKRVLIAAHGNSLRGIVKHLEGMSDQAIMELNLPTGIPIVYELNKELKPTKPMQFLGDEETVRKAMEAVAAQGKAK | |
RUO | |
PGAM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |