missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PEX6 Partial ORF (NP_000278, 881 a.a. - 980 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005190-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102625

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6. [provided by RefSeq]

Sequence: RVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAAC
Specifications

Specifications

NP_000278
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAAC
RUO
PEX6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5190
PEX6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PAF-2/PAF2/PXAAA1
PEX6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers