missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PES1 Partial ORF (NP_055118.1, 3 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023481-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104756

  • 528.00€ / 25µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein that is abnormally elevated in malignant tumors of astrocytic origin. It is a strongly conserved gene containing a BRCT domain that is essential for the activity of this gene product. The gene plays a crucial role in cell proliferation and may be necessary for oncogenic transformation and tumor progression. [provided by RefSeq]

Sequence: GLEKKKYERGSATNYITRNKARKKLQLSLADFRRLCILKGIYPHEPKHKKKVNKGSTAARTFYLIKDIRFLLHEPIVNKFREYKVFVRKLRKAYGKSE
Specifications

Specifications

NP_055118.1
Liquid
23481
PES1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PES
PES1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GLEKKKYERGSATNYITRNKARKKLQLSLADFRRLCILKGIYPHEPKHKKKVNKGSTAARTFYLIKDIRFLLHEPIVNKFREYKVFVRKLRKAYGKSE
RUO
PES1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers