missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PDIK1L Partial ORF (NP_690048, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00149420-Q01.10ug
This item is not returnable.
View return policy
Description
Sequence: MVSSQPKYDLIREVGRGSYGVVYEAVIRKTSARVAVKKIRCHAPENVELALREFWALSSIKSQHPNVIHLEECILQKDGMVQKMSHGSNSSLYLQLVETSpecifications
NP_690048 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVSSQPKYDLIREVGRGSYGVVYEAVIRKTSARVAVKKIRCHAPENVELALREFWALSSIKSQHPNVIHLEECILQKDGMVQKMSHGSNSSLYLQLVET | |
RUO | |
PDIK1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
149420 | |
PDIK1L (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLIK1L/RP11-96L14.4 | |
PDIK1L | |
Recombinant | |
wheat germ expression system |