Learn More
R&D Systems™ Human PDGF-DD Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
Description
The Human PDGF-DD Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PDGF-DD in cell culture supernates (50 ul), serum (10 ul), platelet-poor edta plasma (50 ul), platelet-poor heparin plasma (50 ul), saliva (50 ul), human milk (50 ul).
Specifications
Specifications
| Assay Range | 31.30 - 2000pg/mL |
| Assay Sensitivity | 3.51pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Human Milk, Platelet-poor Plasma (EDTA, Heparin), Saliva, Serum |
| Specificity | Natural and Recombinant Human PDGF-DD |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 80310 |
| Sample Volume | Cell Culture Supernates (50uL),Serum (10uL),Platelet-poor EDTA Plasma (50uL),Platelet-poor Heparin Plasma (50uL),Saliva (50uL),Human Milk (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |