Learn More
Abnova™ Human PDCD2 Full-length ORF (NP_002589.2, 1 a.a. - 344 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005134-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear protein expressed in a variety of tissues. The rat homolog, Rp8, is transiently expressed in immature thymocytes and is thought to be involved in programmed cell death. Expression of the human gene has been shown to be repressed by BCL6, a transcriptional repressor required for lymph node germinal center development, suggesting that BCL6 regulates apoptosis by its effects on PDCD2. This gene is closely linked on chromosome 6 to the gene for TBP, the TATA binding protein. Two transcripts encoding different proteins have been identified. [provided by RefSeq]
Sequence: MAAAGARPVELGFAESAPAWRLRSEQFPSKVGGRPAWLGAAGLPGPQALACELCGRPLSFLLQVYAPLPGRPDAFHRCIFLFCCREQPCCAGLRVFRNQLPRKNDFYSYEPPSENPPPETGESVCLQLKSGAHLCRVCGCLGPKTCSRCHKAYYCSKEHQTLDWRLGHKQACAQPDHLDHIIPDHNFLFPEFEIVIETEDEIMPEVVEKEDYSEIIGSMGEALEEELDSMAKHESREDKIFQKFKTQIALEPEQILRYGRGIAPIWISGENIPQEKDIPDCPCGAKRILEFQVMPQLLNYLKADRLGKSIDWGILAVFTCAESCSLGTGYTEEFVWKQDVTDTPSpecifications
NP_002589.2 | |
Liquid | |
5134 | |
PDCD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAGARPVELGFAESAPAWRLRSEQFPSKVGGRPAWLGAAGLPGPQALACELCGRPLSFLLQVYAPLPGRPDAFHRCIFLFCCREQPCCAGLRVFRNQLPRKNDFYSYEPPSENPPPETGESVCLQLKSGAHLCRVCGCLGPKTCSRCHKAYYCSKEHQTLDWRLGHKQACAQPDHLDHIIPDHNFLFPEFEIVIETEDEIMPEVVEKEDYSEIIGSMGEALEEELDSMAKHESREDKIFQKFKTQIALEPEQILRYGRGIAPIWISGENIPQEKDIPDCPCGAKRILEFQVMPQLLNYLKADRLGKSIDWGILAVFTCAESCSLGTGYTEEFVWKQDVTDTP | |
RUO | |
PDCD2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12347/RP8/ZMYND7 | |
PDCD2 | |
Yes | |
wheat germ expression system |