missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCSK7 Full-length ORF (AAH10696.1, 1 a.a. - 785 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009159-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193342

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease. It is structurally related to its family members, PACE and PACE4. This protein is concentrated in the trans-Golgi network, associated with the membranes, and is not secreted. It can process proalbumin and is thought to be responsible for the activation of HIV envelope glycoproteins gp160 and gp140. This gene has been implicated in the transcriptional regulation of housekeeping genes. Multiple alternatively spliced transcripts are described for this gene but their full length nature is not yet known. Downstream of this gene's map location at 11q23-q24, nucleotides that match part of this gene's 3' end are duplicated and inverted. A translocation breakpoint associated with lymphoma occurs between this gene and its inverted counterpart. [provided by RefSeq]

Sequence: MPKGRQKVPHLDAPLGLPTCLWLELAGLFLLVPWVMGLAGTGGPDGQGTGGPSWAVHLESLEGDGEEETLEQQADALAQAAGLVNAGRIGELQGHYLFVQPAGHRPALEVEAIRQQVEAVLAGHEAVRWHSEQRLLRRAKRSVHFNDPKYPQQWHLNNRRSPGRDINVTGVWERNVTGRGVTVVVVDDGVEHTIQDIAPNYSPEGSYDLNSNDPDPMPHPDVENGNHHGTRCAGEIAAVPNNSFCAVGVAYGSRIAGIRVLDGPLTDSMEAVAFNKHYQINDIYSCSWGPDDDGKTVDGPHQLGKAALQHGVIAGRQGFGSIFVVASGNGGQHNDNCNYDGYANSIYTVTIGAVDEEGRMPFYAEECASMLAVTFSGGDKMLRSIVTTDWDLQKGTGCTEGHTGTSAAAPLAAGMIALMLQVRPCLTWRDVQHIIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLELKLFCPSGMMSLIGAPRSMDSDPNGFNDWTFSTVRCWGERARGTYRLVIRDVGDESFQVGILRQWQLTLYGSVWSAVDIRDRQRLLESAMSGKYLHDDFALPCPPGLKIPEEDGYTITPNTLKTLVLVGCFTVFWTVYYMLEVYLSQRNVASNQVCRSGPCHWPHRSRKAKEEGTELESVPLCSSKDPDEVETESRGPPTTSDLLAPDLLEQGDWSLSQNKSALDCPHQHLDVPHGKEEQIC
Specifications

Specifications

AAH10696.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
111.87kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LPC/PC7/PC8/SPC7
PCSK7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9159
PCSK7 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPKGRQKVPHLDAPLGLPTCLWLELAGLFLLVPWVMGLAGTGGPDGQGTGGPSWAVHLESLEGDGEEETLEQQADALAQAAGLVNAGRIGELQGHYLFVQPAGHRPALEVEAIRQQVEAVLAGHEAVRWHSEQRLLRRAKRSVHFNDPKYPQQWHLNNRRSPGRDINVTGVWERNVTGRGVTVVVVDDGVEHTIQDIAPNYSPEGSYDLNSNDPDPMPHPDVENGNHHGTRCAGEIAAVPNNSFCAVGVAYGSRIAGIRVLDGPLTDSMEAVAFNKHYQINDIYSCSWGPDDDGKTVDGPHQLGKAALQHGVIAGRQGFGSIFVVASGNGGQHNDNCNYDGYANSIYTVTIGAVDEEGRMPFYAEECASMLAVTFSGGDKMLRSIVTTDWDLQKGTGCTEGHTGTSAAAPLAAGMIALMLQVRPCLTWRDVQHIIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLELKLFCPSGMMSLIGAPRSMDSDPNGFNDWTFSTVRCWGERARGTYRLVIRDVGDESFQVGILRQWQLTLYGSVWSAVDIRDRQRLLESAMSGKYLHDDFALPCPPGLKIPEEDGYTITPNTLKTLVLVGCFTVFWTVYYMLEVYLSQRNVASNQVCRSGPCHWPHRSRKAKEEGTELESVPLCSSKDPDEVETESRGPPTTSDLLAPDLLEQGDWSLSQNKSALDCPHQHLDVPHGKEEQIC
RUO
PCSK7
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers