missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCGF5 Full-length ORF (NP_115749.2, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084333-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125853

  • 359.00€ / 10µg

Estimated Shipment: 06-10-2022
to see stock.
Add to basket

Description

Description

Sequence: MATQRKHLVKDFNPYITCYICKGYLIKPTTVTECLHTFCKTCIVQHFEDSNDCPRCGNQVHETNPLEMLRLDNTLEEIIFKLVPGLREQELERESEFWKKNKPQENGQDDTSKADKPKVDEEGDENEDDKDYHRSDPQIAICLDCLRNNGQSGDNVVKGLMKKFIRCSTRVTVGTIKKFLSLKLKLPSSYELDVLCNGEIMGKDHTMEFIYMTRWRLRGENFRCLNCSASQVCSQDGPLYQSYPMVLQYRPRIDFG
Specifications

Specifications

NP_115749.2
Liquid
84333
PCGF5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATQRKHLVKDFNPYITCYICKGYLIKPTTVTECLHTFCKTCIVQHFEDSNDCPRCGNQVHETNPLEMLRLDNTLEEIIFKLVPGLREQELERESEFWKKNKPQENGQDDTSKADKPKVDEEGDENEDDKDYHRSDPQIAICLDCLRNNGQSGDNVVKGLMKKFIRCSTRVTVGTIKKFLSLKLKLPSSYELDVLCNGEIMGKDHTMEFIYMTRWRLRGENFRCLNCSASQVCSQDGPLYQSYPMVLQYRPRIDFG
RUO
PCGF5
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC16202/RNF159
PCGF5
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers