Learn More
Abnova™ Human PCDHB12 Full-length ORF (NP_061755.1, 1 a.a. - 795 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056124-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]
Sequence: MENGGAGTLQIRQVLLFFVLLGMSQAGSETGNFLVMEELQSGSFVGNLAKTLGLEVSELSSRGARVVSNDNKECLQLDTNTGDLLLREMLDREELCGSNEPCVLYFQVLMKNPTQFLQIELQVRDINDHSPVFLEKEMLLEIPENSPVGAVFLLESAKDLDVGINAVKSYTINPNSHFHVKIRVNPDNRKYPELVLDKALDYEERPELSFILTALDGGSPPRSGTALVRVVVVDINDNSPEFEQAFYEVKILENSILGSLVVTVSAWDLDSGTNSELSYTFSHASEDIRKTFEINQKSGDITLTAPLDFEAIESYSIIIQATDGGGLFGKSTVRIQVMDVNDNAPEITVSSITSPIPENTPETVVMVFRIRDRDSGDNGKMVCSIPEDIPFVLKSSVNNYYTLETERPLDRESRAEYNITITVTDLGTPRLKTEHNITVLVSDVNDNAPAFTQTSYALFVRENNSPALHIGSISATDRDSGTNAQVNYSLLPSQDPHLPLASLVSINADNGHLFALRSLDYEALQGFQFRVGATDHGSPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPWAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDAAKHRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAAPVGRCSVPEGPFPGHLVDVSGTGTLSQSYHYEVCVTGGSRSNKFKFLKPIIPNFLPQSTGSEVEENPPFQNNLGFSpecifications
NP_061755.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
113.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCDH-BETA12 | |
PCDHB12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56124 | |
PCDHB12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MENGGAGTLQIRQVLLFFVLLGMSQAGSETGNFLVMEELQSGSFVGNLAKTLGLEVSELSSRGARVVSNDNKECLQLDTNTGDLLLREMLDREELCGSNEPCVLYFQVLMKNPTQFLQIELQVRDINDHSPVFLEKEMLLEIPENSPVGAVFLLESAKDLDVGINAVKSYTINPNSHFHVKIRVNPDNRKYPELVLDKALDYEERPELSFILTALDGGSPPRSGTALVRVVVVDINDNSPEFEQAFYEVKILENSILGSLVVTVSAWDLDSGTNSELSYTFSHASEDIRKTFEINQKSGDITLTAPLDFEAIESYSIIIQATDGGGLFGKSTVRIQVMDVNDNAPEITVSSITSPIPENTPETVVMVFRIRDRDSGDNGKMVCSIPEDIPFVLKSSVNNYYTLETERPLDRESRAEYNITITVTDLGTPRLKTEHNITVLVSDVNDNAPAFTQTSYALFVRENNSPALHIGSISATDRDSGTNAQVNYSLLPSQDPHLPLASLVSINADNGHLFALRSLDYEALQGFQFRVGATDHGSPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPWAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDAAKHRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAAPVGRCSVPEGPFPGHLVDVSGTGTLSQSYHYEVCVTGGSRSNKFKFLKPIIPNFLPQSTGSEVEENPPFQNNLGF | |
RUO | |
PCDHB12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |