missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PATZ1 Partial ORF (NP_114440.1, 260 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023598-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194886

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene contains an A-T hook DNA binding motif which usually binds to other DNA binding structures to play an important role in chromatin modeling and transcription regulation. Its Poz domain is thought to function as a site for protein-protein interaction and is required for transcriptional repression, and the zinc-fingers comprise the DNA binding domain. Since the encoded protein has typical features of a transcription factor, it is postulated to be a repressor of gene expression. In small round cell sarcoma, this gene is fused to EWS by a small inversion of 22q, then the hybrid is thought to be translocated (t(1;22)(p36.1;q12). The rearrangement of chromosome 22 involves intron 8 of EWS and exon 1 of this gene creating a chimeric sequence containing the transactivation domain of EWS fused to zinc finger domain of this protein. This is a distinct example of an intra-chromosomal rearrangement of chromosome 22. Four alternatively spliced transcript variants are described for this gene. [provided by RefSeq]

Sequence: PLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSRTRKQVACE
Specifications

Specifications

NP_114440.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSRTRKQVACE
RUO
PATZ1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23598
PATZ1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAZR/PATZ/RIAZ/ZBTB19/ZNF278/ZSG/dJ400N23
PATZ1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers