missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PASD1 Partial ORF (NP_775764.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00139135-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16095775

  • 335.00€ / 10µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MKMRGEKRRDKVNPKSSQRKLNWIPSFPTYDYFNQVTLQLLDGFMITLSTDGVIICVAENISSLLGHLPAEIVGKKLLSLLPDEEKDEVYQKIILKFPLL
Specifications

Specifications

NP_775764.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKMRGEKRRDKVNPKSSQRKLNWIPSFPTYDYFNQVTLQLLDGFMITLSTDGVIICVAENISSLLGHLPAEIVGKKLLSLLPDEEKDEVYQKIILKFPLL
RUO
PASD1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
139135
PASD1 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PASD1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers