missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PARC Partial ORF (NP_055904, 918 a.a. - 1025 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023113-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MMLLNRYSEPPGSPERAALETPIIQGQDGSPELLIRSLVGGPSAELLLDLERVLCREGSPGGAVRPLLKRLQQETQPFLLLLRTLDAPGPNKTLLLSVLRVITRLLDFSpecifications
NP_055904 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMLLNRYSEPPGSPERAALETPIIQGQDGSPELLIRSLVGGPSAELLLDLERVLCREGSPGGAVRPLLKRLQQETQPFLLLLRTLDAPGPNKTLLLSVLRVITRLLDF | |
RUO | |
CUL9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23113 | |
PARC (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686G1042/DKFZp686P2024/H7AP1/PARC/RP3-330M21.2 | |
CUL9 | |
Recombinant | |
wheat germ expression system |