missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PAN3 Partial ORF (NP_787050, 508 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00255967-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: ENLQKAMELVTINYSSDLKNLILYLLTDQNRMRSVNDIMPMIGARFYTQLDAAQMRNDVIEEDLAKEVQNGRLFRLLAKLGTINERPEFQKDPTWSETGDRYLLKLFRSpecifications
NP_787050 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ENLQKAMELVTINYSSDLKNLILYLLTDQNRMRSVNDIMPMIGARFYTQLDAAQMRNDVIEEDLAKEVQNGRLFRLLAKLGTINERPEFQKDPTWSETGDRYLLKLFR | |
RUO | |
PAN3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
255967 | |
PAN3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |