Learn More
Abnova™ Human PAGE1 Full-length ORF (NP_003776.2, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008712-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. Unlike the other gene family members, this gene does not encode an antigenic peptide. Nothing is presently known about the function of this protein. [provided by RefSeq]
Sequence: MGFLRRLIYRRRPMIYVESSEESSDEQPDEVESPTQSQDSTPAEEREDEGASAAQGQEPEADSQELVQPKTGCELGDGPDTKRVCLRNEEQMKLPAEGPEPEADSQEQVHPKTGCERGDGPDVQELGLPNPEEVKTPEEDEGQSQPSpecifications
NP_003776.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAGE-9/GAGEB1/PAGE-1 | |
PAGE1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8712 | |
PAGE1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGFLRRLIYRRRPMIYVESSEESSDEQPDEVESPTQSQDSTPAEEREDEGASAAQGQEPEADSQELVQPKTGCELGDGPDTKRVCLRNEEQMKLPAEGPEPEADSQEQVHPKTGCERGDGPDVQELGLPNPEEVKTPEEDEGQSQP | |
RUO | |
PAGE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |