missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PADI1 Partial ORF (NP_037490.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029943-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156166

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type I enzyme is involved in the late stages of epidermal differentiation, where it deiminates filaggrin and keratin K1, which maintains hydration of the stratum corneum, and hence the cutaneous barrier function. This enzyme may also play a role in hair follicle formation. This gene exists in a cluster with four other paralogous genes. [provided by RefSeq]

Sequence: MAPKRVVQLSLKMPTHAVCVVGVEAHVDIHSDVPKGANSFRVSGSSGVEVFMVYNRTRVKEPIGKARWPLDTDADMVVSVGTASKELKDFKVRVSYFGEQ
Specifications

Specifications

NP_037490.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAPKRVVQLSLKMPTHAVCVVGVEAHVDIHSDVPKGANSFRVSGSSGVEVFMVYNRTRVKEPIGKARWPLDTDADMVVSVGTASKELKDFKVRVSYFGEQ
RUO
PADI1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
29943
PADI1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPAD10/PAD1/PDI/PDI1
PADI1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers