missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human P2RX2 Partial ORF (NP_036358.2, 128 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022953-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194296

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: KNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEKAGESFTELAHKGGVIGVIINWDCDLDLPAS
Specifications

Specifications

NP_036358.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.32kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEKAGESFTELAHKGGVIGVIINWDCDLDLPAS
RUO
P2RX2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
22953
P2RX2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC129601/P2X2
P2RX2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers