missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human P-Selectin/CD62P Quantikine ELISA Kit

Product Code. 18742124
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18742124

Brand: R&D Systems™ SPSE00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human P-Selectin/CD62P Quantikine ELISA Kit quantifies human P-Selectin/CD62P in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul).

1 Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.78 - 50 ng/mL
Assay Sensitivity 0.121 ng/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human P-Selectin
For Use With (Application) ELISA
Gene ID (Entrez) 6403
Host Species Human
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (10 uL),Serum (10 uL),EDTA Plasma (10 uL),Heparin Plasma (10 uL),Citrate Plasma (10 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140 kDa, antigen CD62), SELP
Target P-Selectin/CD62P
Total Assay Time 1.25 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.