missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OXCT2 Full-length ORF (AAH30015.1, 1 a.a. - 513 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064064-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16163343

  • 508.00€ / 25µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

OXCT2 is a testis-specific succinyl-CoA:3-oxoacid CoA transferase (EC 2.8.3.5), which catalyzes the reversible transfer of CoA from succinyl-CoA to acetoacetate in the first step of ketone body utilization. See also OXCT1 (MIM 601424).[supplied by OMIM]

Sequence: MAALRLLASVLGRGVPAGGSGLALSQGCARCFATSPRLRAKFYADPVEMVKDISDGATVMIGGFGLCGIPENLIAALLRTRVKDLQVVSSNVGVEDFGLGLLLAARQVRRIVCSYVGENTLCESQYLAGELELELTPQGTLAERIRAGGAGVPAFYTPTGYGTLVQEGGAPIRYTPDGHLALMSQPREVREFNGDHFLLERAIRADFALVKGWKADRAGNVVFRRSARNFNVPMCKAADVTAVEVEEIVEVGAFPPEDIHVPNIYVDRVIKGQKYEKRIERLTILKEEDGDAGKEEDARTRIIRRAALEFEDGMYANLGIGIPLLASNFISPSMTVHLHSENGILGLGPFPTEDEVDADLINAGKQTVTVLPGGCFFASDDSFAMIRGGHIQLTMLGAMQVSKYGDLANWMIPGKKVKGMGGAMDLVSSQKTRVVVTMQHCTKDNTPKIMEKCTMPLTGKRCVDRIITEKAVFDVHRKKELTLRELWEGLTVDDIKKSTGCAFAVSPNLRPMQ
Specifications

Specifications

AAH30015.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
82.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FKSG25/FLJ00030/SCOT-T
OXCT2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64064
OXCT2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAALRLLASVLGRGVPAGGSGLALSQGCARCFATSPRLRAKFYADPVEMVKDISDGATVMIGGFGLCGIPENLIAALLRTRVKDLQVVSSNVGVEDFGLGLLLAARQVRRIVCSYVGENTLCESQYLAGELELELTPQGTLAERIRAGGAGVPAFYTPTGYGTLVQEGGAPIRYTPDGHLALMSQPREVREFNGDHFLLERAIRADFALVKGWKADRAGNVVFRRSARNFNVPMCKAADVTAVEVEEIVEVGAFPPEDIHVPNIYVDRVIKGQKYEKRIERLTILKEEDGDAGKEEDARTRIIRRAALEFEDGMYANLGIGIPLLASNFISPSMTVHLHSENGILGLGPFPTEDEVDADLINAGKQTVTVLPGGCFFASDDSFAMIRGGHIQLTMLGAMQVSKYGDLANWMIPGKKVKGMGGAMDLVSSQKTRVVVTMQHCTKDNTPKIMEKCTMPLTGKRCVDRIITEKAVFDVHRKKELTLRELWEGLTVDDIKKSTGCAFAVSPNLRPMQ
RUO
OXCT2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers