missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OVGP1 Partial ORF (NP_002548.3, 579 a.a. - 677 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005016-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102265

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a large, carbohydrate-rich, epithelial glycoprotein with numerous O-glycosylation sites located within threonine, serine, and proline-rich tandem repeats. The gene is similar to members of the mucin and the glycosyl hydrolase 18 gene families. Regulation of expression may be estrogen-dependent. Gene expression and protein secretion occur during late follicular development through early cleavage-stage embryonic development. The protein is secreted from non-ciliated oviductal epithelial cells and associates with ovulated oocytes, blastomeres, and spermatozoan acrosomal regions. [provided by RefSeq]

Sequence: RNISVTPEGQTMPLRGENLTSEVGTHPRMGNLGLQMEAENRMMLSSSPVIQLPEQTPLAFDNRFVPIYGNHSSVNSVTPQTSPLSLKKEIPENSAVDEE
Specifications

Specifications

NP_002548.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RNISVTPEGQTMPLRGENLTSEVGTHPRMGNLGLQMEAENRMMLSSSPVIQLPEQTPLAFDNRFVPIYGNHSSVNSVTPQTSPLSLKKEIPENSAVDEE
RUO
OVGP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5016
OVGP1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CHIT5/EGP/MUC9/OGP
OVGP1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers