Learn More
Abnova™ Human OTOS Partial ORF (NP_683764.1, 26 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00150677-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Otospiralin is synthesized by nonsensory cells (fibrocytes) of the inner ear, and downregulation of otospiralin in guinea pigs leads to deafness (Lavigne-Rebillard et al., 2003 [PubMed 12687421]).[supplied by OMIM]
Sequence: EEGDPYAELPAMPYWPFSTSDFWNYVQHFQALGAYPQIEDMARTFFAHFPLGSTLGFHVPYQEDSpecifications
NP_683764.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.78kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEGDPYAELPAMPYWPFSTSDFWNYVQHFQALGAYPQIEDMARTFFAHFPLGSTLGFHVPYQED | |
RUO | |
OTOS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
150677 | |
OTOS (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OTOSP | |
OTOS | |
Recombinant | |
wheat germ expression system |