Learn More
Abnova™ Human OTOA Partial ORF (NP_653273.3, 27 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_653273.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 146183 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132037
|
Abnova™
H00146183-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16122037
|
Abnova™
H00146183-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is specifically expressed in the inner ear, and is located at the interface between the apical surface of the inner ear sensory epithelia and their overlying acellular gels. It is prposed that this protein is involved in the attachment of the inner ear acellular gels to the apical surface of the underlying nonsensory cells. Mutations in this gene are associated with autosomal recessive deafness type 22 (DFNB22). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: NSRQDLHPLLQNMAEEIIDGSYLNALLDLIQFQSSHVWTDDLSHRVLAYLNSRNVAFTIPSLQAAVENHLEQRLHQPQKLLEDLRKTDAQQFRTAMKCSpecifications
NP_653273.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB22/FLJ32773/MGC157747/MGC39813 | |
OTOA | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
146183 | |
OTOA (Human) Recombinant Protein (Q01) | |
NSRQDLHPLLQNMAEEIIDGSYLNALLDLIQFQSSHVWTDDLSHRVLAYLNSRNVAFTIPSLQAAVENHLEQRLHQPQKLLEDLRKTDAQQFRTAMKC | |
RUO | |
OTOA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |