missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Osteopontin (OPN) Quantikine ELISA Kit

Product Code. 18792014
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18792014

Brand: R&D Systems™ SOST00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Osteopontin (OPN) Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Osteopontin (OPN) Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Osteopontin/OPN in cell culture supernates (50 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul), human milk (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 - 20ng/mL
Assay Sensitivity 0.024ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Urine
Specificity Natural and Recombinant Human Osteopontin
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 6696
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1 bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.