missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBPL6 Full-length ORF (AAH52259.1, 1 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00114880-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107143

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MSSDEKGISPAHKTSTPTHRSASSSTSSQRDSRQSIHILERTASSSTEPSVSRQLLEPEPVPLSKEADSWEIIEGLKIGQTNVQKPDKHEGFMLKKRKWPLKGWHKRFFVLDNGMLKYSKAPLDIQKGKVHGSIDVGLSVMSIKKKARRIDLDTEEHIYHLKVKSQDWFDAWVSKLRHHRLYRQNEIVRSPRDASFHIFPSTSTAESSPAANVSVMDGKMQPNSFPWQSPLPCSNSLPATCTTGQSKVAAWLQDSEEMDRCAEDLAHCQSNLVELSKLLQNLEILQRTQSAPNFTDMQVPFSATMSPVRLHSSNPNLCADIEFQTPPSHLTDPLESSTDYTKLQEEFCLIAQKVHSLLKSAFNSIAIEKEKLKQMVSEQDHSKGHSTQMARLRQSLSQAGEQIHVSLPLSQQVANESRLSMSESVSEFFDAQEVLLSASSSENEASDDESYISDVSDNISEDNTSVADNISRQSMHHLSFQVVLSTCQIATRRLNEQAFPLPRHPHPC
Specifications

Specifications

AAH52259.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ36583/MGC59642/ORP6
OSBPL6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
114880
OSBPL6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSDEKGISPAHKTSTPTHRSASSSTSSQRDSRQSIHILERTASSSTEPSVSRQLLEPEPVPLSKEADSWEIIEGLKIGQTNVQKPDKHEGFMLKKRKWPLKGWHKRFFVLDNGMLKYSKAPLDIQKGKVHGSIDVGLSVMSIKKKARRIDLDTEEHIYHLKVKSQDWFDAWVSKLRHHRLYRQNEIVRSPRDASFHIFPSTSTAESSPAANVSVMDGKMQPNSFPWQSPLPCSNSLPATCTTGQSKVAAWLQDSEEMDRCAEDLAHCQSNLVELSKLLQNLEILQRTQSAPNFTDMQVPFSATMSPVRLHSSNPNLCADIEFQTPPSHLTDPLESSTDYTKLQEEFCLIAQKVHSLLKSAFNSIAIEKEKLKQMVSEQDHSKGHSTQMARLRQSLSQAGEQIHVSLPLSQQVANESRLSMSESVSEFFDAQEVLLSASSSENEASDDESYISDVSDNISEDNTSVADNISRQSMHHLSFQVVLSTCQIATRRLNEQAFPLPRHPHPC
RUO
OSBPL6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers