Learn More
Abnova™ Human OSBPL3 Full-length ORF (NP_663162.1, 1 a.a. - 820 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026031-P01.10ug
Detalles adicionales : Weight : 0.00010kg
Descripción
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Several transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MMSDEKNLGVSQKLVSPSRSTSSCSSKQGSRQDSWEVVEGLRGEMNYTQEPPVQKGFLLKKRKWPLKGWHKRFFYLDKGILKYAKSQTDIEREKLHGCIDVGLSVMSVKKSSKCIDLDTEEHIYHLKVKSEEVFDEWVSKLRHHRMYRQNEIAMFPHEVNHFFSGSTITDSSSGVFDSISSRKRSSISKQNLFQTGSNVSFSCGGETRVPLWLQSSEDMEKCSKDLAHCHAYLVEMSQLLQSMDVLHRTYSAPAINAIQVPKPFSGPVRLHSSNPNLSTLDFGEEKNYSDGSETSSEFSKMQEDLCHIAHKVYFTLRSAFNIMSAEREKLKQLMEQDASSSPSAQVIGLKNALSSENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPCPSSSNISLWNILRNNIGKDLSKVAMPVELNEPLNTLQRLCEELEYSELLDKAAQIPSPLERMVYVAAFAISAYASSYYRAGSKPFNPVLGETYECIREDKGFQFFSEQVSHHPPISACHAESRNFVFWQDVRWKNKFWGKSMEIVPIGTTHVTLPVFGDHFEWNKVTSCIHNILSGQRWIEHYGEIVIKNLHDDSCYCKVNFIKAKYWSTNAHEIEGTVFDRSGKAVHRLFGKWHESIYCGGGSSSACVWRANPMPKGYEQYYSFTQFALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLWEspecificaciones
NP_663162.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
120.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp667P1518/KIAA0704/MGC21526/ORP-3/ORP3/OSBP3 | |
OSBPL3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26031 | |
OSBPL3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMSDEKNLGVSQKLVSPSRSTSSCSSKQGSRQDSWEVVEGLRGEMNYTQEPPVQKGFLLKKRKWPLKGWHKRFFYLDKGILKYAKSQTDIEREKLHGCIDVGLSVMSVKKSSKCIDLDTEEHIYHLKVKSEEVFDEWVSKLRHHRMYRQNEIAMFPHEVNHFFSGSTITDSSSGVFDSISSRKRSSISKQNLFQTGSNVSFSCGGETRVPLWLQSSEDMEKCSKDLAHCHAYLVEMSQLLQSMDVLHRTYSAPAINAIQVPKPFSGPVRLHSSNPNLSTLDFGEEKNYSDGSETSSEFSKMQEDLCHIAHKVYFTLRSAFNIMSAEREKLKQLMEQDASSSPSAQVIGLKNALSSENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPCPSSSNISLWNILRNNIGKDLSKVAMPVELNEPLNTLQRLCEELEYSELLDKAAQIPSPLERMVYVAAFAISAYASSYYRAGSKPFNPVLGETYECIREDKGFQFFSEQVSHHPPISACHAESRNFVFWQDVRWKNKFWGKSMEIVPIGTTHVTLPVFGDHFEWNKVTSCIHNILSGQRWIEHYGEIVIKNLHDDSCYCKVNFIKAKYWSTNAHEIEGTVFDRSGKAVHRLFGKWHESIYCGGGSSSACVWRANPMPKGYEQYYSFTQFALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLW | |
RUO | |
OSBPL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |