missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR3A3 Full-length ORF (NP_036505.1, 1 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008392-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102412

  • 528.00€ / 25µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MSLQKLMEPEAGTNRTAVAEFILLGLVQTEEMQPVVFVLLLFAYLVTTGGNLSILAAVLVEPKLHAPMYFFLGNLSVLDVGCITVTVPAMLGRLLSHKSTISYDACLSQLFFFHLLAGMDCFLLTAMAYDRLLAICQPLTYSTRMSQTVQRMLVAASWACAFTNALTHTVAMSTLNFCGPNEVNHFYCDLPQLFQLSCSSTQLNELLLFVAAAFMAVAPLVFISVSYAHVVAAVLQIRSAEGRKKAFSTCGSHLTVVGIFYGTGVFSYMRLGSVESSDKDKGVGVFMTVINPMLNPLIYSLRNTDVQGALCQLLVGERSLT
Specifications

Specifications

NP_036505.1
Liquid
8392
OR3A3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLQKLMEPEAGTNRTAVAEFILLGLVQTEEMQPVVFVLLLFAYLVTTGGNLSILAAVLVEPKLHAPMYFFLGNLSVLDVGCITVTVPAMLGRLLSHKSTISYDACLSQLFFFHLLAGMDCFLLTAMAYDRLLAICQPLTYSTRMSQTVQRMLVAASWACAFTNALTHTVAMSTLNFCGPNEVNHFYCDLPQLFQLSCSSTQLNELLLFVAAAFMAVAPLVFISVSYAHVVAAVLQIRSAEGRKKAFSTCGSHLTVVGIFYGTGVFSYMRLGSVESSDKDKGVGVFMTVINPMLNPLIYSLRNTDVQGALCQLLVGERSLT
RUO
OR3A3
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR17-137/OR17-16/OR17-201/OR3A6/OR3A7/OR3A8P
OR3A3
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers