missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR13D1 Partial ORF (NP_001004484, 2 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00286365-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192767

  • 528.00€ / 25µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: YRFTDFDVSNISIYLNHVLFYTTQQAGDLEHMETRNYSAMTEFFLVGLSQYPELQ
Specifications

Specifications

NP_001004484
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.79kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YRFTDFDVSNISIYLNHVLFYTTQQAGDLEHMETRNYSAMTEFFLVGLSQYPELQ
RUO
OR13D1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
286365
OR13D1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR9-15
OR13D1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers