missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OLFM3 Partial ORF (NP_477518, 108 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00118427-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: FRQIEDDRKTLMTKHFQELKEKMDELLPLIPVLEQYKTDAKLITQFKEEIRNLSAVLTGIQEEIGAYDYEELHQRVLSLETRLRDCMKKLTCGKLMKITSpecifications
NP_477518 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FRQIEDDRKTLMTKHFQELKEKMDELLPLIPVLEQYKTDAKLITQFKEEIRNLSAVLTGIQEEIGAYDYEELHQRVLSLETRLRDCMKKLTCGKLMKIT | |
RUO | |
OLFM3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
118427 | |
OLFM3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NOE3/NOELIN3/NOELIN3_V1/NOELIN3_V2/NOELIN3_V3/NOELIN3_V4/NOELIN3_V5/NOELIN3_V6/OPTIMEDIN | |
OLFM3 | |
Recombinant | |
wheat germ expression system |