missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NSFL1C Full-length ORF (NP_057227.2, 1 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055968-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16171993

  • 508.00€ / 25µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

N-ethylmaleimide-sensitive factor (NSF) and valosin-containing protein (p97) are two ATPases known to be involved in transport vesicle/target membrane fusion and fusions between membrane compartments. A trimer of the protein encoded by this gene binds a hexamer of cytosolic p97 and is required for p97-mediated regrowth of Golgi cisternae from mitotic Golgi fragments. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAAERQEALREFVAVTGAEEDRARFFLESAGWDLQIALASFYEDGGDEDIVTISQATPSSVSRGTAPSDNRVTSFRDLIHDQDEDEEEEEGQRFYAGGSERSGQQIVGPPRKKSPNELVDDLFKGAKEHGAVAVERVTKSPGETSKPRPFAGGGYRLGAAPEEESAYVAGEKRQHSSQDVHVVLKLWKSGFSLDNGELRSYQDPSNAQFLESIRRGEVPAELRRLAHGGQVNLDMEDHRDEDFVKPKGAFKAFTGEGQKLGSTAPQVLSTSSPAQQAENEAKASSSILIDESEPTTNIQIRLADGGRLVQKFNHSHRISDIRLFIVDARPAMAATSFILMTTFPNKELADESQTLKEANLLNAVIVQRLT
Specifications

Specifications

NP_057227.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC3347/UBX1/UBXD10/UBXN2C/dJ776F14.1/p47
NSFL1C
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55968
NSFL1C (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAERQEALREFVAVTGAEEDRARFFLESAGWDLQIALASFYEDGGDEDIVTISQATPSSVSRGTAPSDNRVTSFRDLIHDQDEDEEEEEGQRFYAGGSERSGQQIVGPPRKKSPNELVDDLFKGAKEHGAVAVERVTKSPGETSKPRPFAGGGYRLGAAPEEESAYVAGEKRQHSSQDVHVVLKLWKSGFSLDNGELRSYQDPSNAQFLESIRRGEVPAELRRLAHGGQVNLDMEDHRDEDFVKPKGAFKAFTGEGQKLGSTAPQVLSTSSPAQQAENEAKASSSILIDESEPTTNIQIRLADGGRLVQKFNHSHRISDIRLFIVDARPAMAATSFILMTTFPNKELADESQTLKEANLLNAVIVQRLT
RUO
NSFL1C
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers