missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NR2C1 Full-length ORF (NP_001027458.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007181-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193778

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: MATIEEIAHQIIEQQMGEIVTEQQTGQKIQIVTALDHNTQGKQFILTNHDGSTPSKVILARQDSTPGKVFLTTPDAAGVNQLFFTTPDLSAQHLQLLTDNSPDQGPNKVFDLCVVCGDKASGRHYGAVTCEGCKGFFKRSIRKNLVYSCRGSKDCIINKHHRNRCQYCRLQRCIAFGMKQDSVQCERKPIEVSREKSSNCAASTEKIYIRKDLRSPLTATPTFVTDSESTRSTGLLDSGMFMNIHPSGVKTESAVLMTSDKAESCQGDLSTLANVVTSLANLGKTKDLSQNSNEMSMIESLSNDDTSLCEFQEMQTNGDVSRAFDTLAKALNPGESTACQSSVAGMEGSVHLITGDSSINYTEKEGPLLSDSHVAFRLTMPSPMPEYLNVHYIGESASRLLFLSMHWALSIPSFQALGQENSISLVKAYWNELFTLGLAQCWQVMNVATILATFVNCLHNSLQQAEG
Specifications

Specifications

NP_001027458.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TR2
NR2C1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7181
NR2C1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATIEEIAHQIIEQQMGEIVTEQQTGQKIQIVTALDHNTQGKQFILTNHDGSTPSKVILARQDSTPGKVFLTTPDAAGVNQLFFTTPDLSAQHLQLLTDNSPDQGPNKVFDLCVVCGDKASGRHYGAVTCEGCKGFFKRSIRKNLVYSCRGSKDCIINKHHRNRCQYCRLQRCIAFGMKQDSVQCERKPIEVSREKSSNCAASTEKIYIRKDLRSPLTATPTFVTDSESTRSTGLLDSGMFMNIHPSGVKTESAVLMTSDKAESCQGDLSTLANVVTSLANLGKTKDLSQNSNEMSMIESLSNDDTSLCEFQEMQTNGDVSRAFDTLAKALNPGESTACQSSVAGMEGSVHLITGDSSINYTEKEGPLLSDSHVAFRLTMPSPMPEYLNVHYIGESASRLLFLSMHWALSIPSFQALGQENSISLVKAYWNELFTLGLAQCWQVMNVATILATFVNCLHNSLQQAEG
RUO
NR2C1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers