missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NPHP4 Full-length ORF (AAH50076.1, 1 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH50076.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 261734 |
Molecular Weight (g/mol) | 34.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16119783
|
Abnova™
H00261734-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16109783
|
Abnova™
H00261734-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein which contains a proline-rich region. The encoded protein may function in renal tubular development and function. This protein interacts with nephrocystin. Mutations in this gene are associated with nephronophthisis type 4. Multiple alternative transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: MCGAFSTQAGAVEKTLFDISPAPGAPVGMLGEDPPVHVRCSDPNVICETQNVGPGEPRDIFLKVASGPSPEIKDFFVIIYSSpecifications
AAH50076.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MCGAFSTQAGAVEKTLFDISPAPGAPVGMLGEDPPVHVRCSDPNVICETQNVGPGEPRDIFLKVASGPSPEIKDFFVIIYS | |
RUO | |
NPHP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
261734 | |
NPHP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0673/SLSN4 | |
NPHP4 | |
Recombinant | |
wheat germ expression system |