missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NOVA1 Full-length ORF (NP_002506.2, 1 a.a. - 507 a.a.) Recombinant Protein MW: 81.29kDa with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004857-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122035

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a neuron-specific RNA-binding protein, a member of the Nova family of paraneoplastic disease antigens, that is recognized and inhibited by paraneoplastic antibodies. These antibodies are found in the sera of patients with paraneoplastic opsoclonus-ataxia, breast cancer, and small cell lung cancer. Alternatively spliced transcripts encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: MMAAAPIQQNGTHTGVPIDLDPPDSRKRPLEAPPEAGSTKRTNTGEDGQYFLKVLIPSYAAGSIIGKGGQTIVQLQKETGATIKLSKSKDFYPGTTERVCLIQGTVEALNAVHGFIAEKIREMPQNVAKTEPVSILQPQTTVNPDRIKQTLPSSPTTTKSSPSDPMTTSRANQVKIIVPNSTAGLIIGKGGATVKAVMEQSGAWVQLSQKPDGINLQERVVTVSGEPEQNRKAVELIIQKIQEDPQSGSCLNISYANVTGPVANSNPTGSPYANTAEVLPTAAAAAGLLGHANLAGVAAFPAVLSGFTGNDLVAITSALNTLASYGYNLNTLGLGLSQAAATGALAAAAASANPAAAAANLLATYASEASASGSTAGGTAGTFALGSLAAATAATNGYFGAASPLAASAILGTEKSTDGSKDVVEIAVPENLVGAILGKGGKTLVEYQELTGARIQISKKGEFVPGTRNRKVTITGTPAATQAAQYLITQRITYEQGVRAANPQKVG
Specifications

Specifications

NP_002506.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
81.29kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Nova-1
NOVA1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4857
NOVA1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMAAAPIQQNGTHTGVPIDLDPPDSRKRPLEAPPEAGSTKRTNTGEDGQYFLKVLIPSYAAGSIIGKGGQTIVQLQKETGATIKLSKSKDFYPGTTERVCLIQGTVEALNAVHGFIAEKIREMPQNVAKTEPVSILQPQTTVNPDRIKQTLPSSPTTTKSSPSDPMTTSRANQVKIIVPNSTAGLIIGKGGATVKAVMEQSGAWVQLSQKPDGINLQERVVTVSGEPEQNRKAVELIIQKIQEDPQSGSCLNISYANVTGPVANSNPTGSPYANTAEVLPTAAAAAGLLGHANLAGVAAFPAVLSGFTGNDLVAITSALNTLASYGYNLNTLGLGLSQAAATGALAAAAASANPAAAAANLLATYASEASASGSTAGGTAGTFALGSLAAATAATNGYFGAASPLAASAILGTEKSTDGSKDVVEIAVPENLVGAILGKGGKTLVEYQELTGARIQISKKGEFVPGTRNRKVTITGTPAATQAAQYLITQRITYEQGVRAANPQKVG
RUO
NOVA1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers