Learn More
Abnova™ Human NOL5A Full-length ORF (AAH04937.1, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010528-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. The protein encoded by this gene is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them has not been determined. [provided by RefSeq]
Sequence: MKEAMVQAEEAAAEITRKLEKQEKKRLKKEKKRLAALALASSENSSSTPEECEEMSEKPKKKKKQKPQEVPQENGMEDPSISFSKPKKKKSFSKEELMSSDLEETAGSTSIPKRKKSTPKEETVNDPEEAGHRSGSKKKRKFSKEEPVSSGPEEAVGKSSSKKKKKFHKASQEDSpecifications
AAH04937.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NOL5A | |
NOP56 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10528 | |
NOL5A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKEAMVQAEEAAAEITRKLEKQEKKRLKKEKKRLAALALASSENSSSTPEECEEMSEKPKKKKKQKPQEVPQENGMEDPSISFSKPKKKKSFSKEELMSSDLEETAGSTSIPKRKKSTPKEETVNDPEEAGHRSGSKKKRKFSKEEPVSSGPEEAVGKSSSKKKKKFHKASQED | |
RUO | |
NOP56 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |