Learn More
Abnova™ Human NLRP13 Full-length ORF (AAI48743.1, 1 a.a. - 1043 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00126204-P01.10ug
Additional Details : Weight : 0.00010kg
Description
NALPs are cytoplasmic proteins that form a subfamily within the larger CATERPILLER protein family. Most short NALPs, such as NALP13, have an N-terminal pyrin (MEFV; MIM 608107) domain (PYD), followed by a NACHT domain, a NACHT-associated domain (NAD), and a C-terminal leucine-rich repeat (LRR) region. The long NALP, NALP1 (MIM 606636), also has a C-terminal extension containing a function to find domain (FIIND) and a caspase recruitment domain (CARD). NALPs are implicated in the activation of proinflammatory caspases (e.g., CASP1; MIM 147678) via their involvement in multiprotein complexes called inflammasomes (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM]
Sequence: MNFSVITCPNGGTNQGLLPYLMALDQYQLEEFKLCLEPQQLMDFWSAPQGHFPRIPWANLRAADPLNLSFLLDEHFPKGQAWKVVLGIFQTMNLTSLCEKVRAEMKENVQTQELQDPTQEDLEMLEAAAGNMQTQGCQDPNQEELDELEEETGNVQAQGCQDPNQEEPEMLEEADHRRKYRENMKAELLETWDNISWPKDHVYIRNTSKDEHEELQRLLDPNRTRAQAQTIVLVGRAGVGKTTLAMQAMLHWANGVLFQQRFSYVFYLSCHKIRYMKETTFAELISLDWPDFDAPIEEFMSQPEKLLFIIDGFEEIIISESRSESLDDGSPCTDWYQELPVTKILHSLLKKELVPLATLLITIKTWFVRDLKASLVNPCFVQITGFTGDDLRVYFMRHFDDSSEVEKILQQLRKNETLFHSCSAPMVCWTVCSCLKQPKVRYYDLQSITQTTTSLYAYFFSNLFSTAEVDLADDSWPGQWRALCSLAIEGLWSMNFTFNKEDTEIEGLEVPFIDSLYEFNILQKINDCGGCTTFTHLSFQEFFAAMSFVLEEPREFPPHSTKPQEMKMLLQHVLLDKEAYWTPVVLFFFGLLNKNIARELEDTLHCKISPRVMEELLKWGEELGKAESASLQFHILRLFHCLHESQEEDFTKKMLGRIFEVDLNILEDEELQASSFCLKHCKRLNKLRLSVSSHILERDLEILETSKFDSRMHAWNSICSTLVTNENLHELDLSNSKLHASSVKGLCLALKNPRCKVQKLTCKSVTPEWVLQDLIIALQGNSKLTHLNFSSNKLGMTVPLILKALRHSACNLKYLCLEKCNLSAASCQDLALFLTSIQHVTRLCLGFNRLQDDGIKLLCAALTHPKCALERLELWFCQLAAPACKHLSDALLQNRSLTHLNLSKNSLRDEGVKFLCEALGRPDGNLQSLNLSGCSFTREGCGELANALSHNHNVKILDLGENDLQDDGVKLLCEALKPHRALHTLGLAKCNLTTACCQHLFSVLSSSKSLVNLNLLGNELDTDGVKMLCKALKKSTCRLQKLGSpecifications
AAI48743.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
141.68kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLR19.7/NALP13/NOD14/PAN13 | |
NLRP13 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
126204 | |
NLRP13 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNFSVITCPNGGTNQGLLPYLMALDQYQLEEFKLCLEPQQLMDFWSAPQGHFPRIPWANLRAADPLNLSFLLDEHFPKGQAWKVVLGIFQTMNLTSLCEKVRAEMKENVQTQELQDPTQEDLEMLEAAAGNMQTQGCQDPNQEELDELEEETGNVQAQGCQDPNQEEPEMLEEADHRRKYRENMKAELLETWDNISWPKDHVYIRNTSKDEHEELQRLLDPNRTRAQAQTIVLVGRAGVGKTTLAMQAMLHWANGVLFQQRFSYVFYLSCHKIRYMKETTFAELISLDWPDFDAPIEEFMSQPEKLLFIIDGFEEIIISESRSESLDDGSPCTDWYQELPVTKILHSLLKKELVPLATLLITIKTWFVRDLKASLVNPCFVQITGFTGDDLRVYFMRHFDDSSEVEKILQQLRKNETLFHSCSAPMVCWTVCSCLKQPKVRYYDLQSITQTTTSLYAYFFSNLFSTAEVDLADDSWPGQWRALCSLAIEGLWSMNFTFNKEDTEIEGLEVPFIDSLYEFNILQKINDCGGCTTFTHLSFQEFFAAMSFVLEEPREFPPHSTKPQEMKMLLQHVLLDKEAYWTPVVLFFFGLLNKNIARELEDTLHCKISPRVMEELLKWGEELGKAESASLQFHILRLFHCLHESQEEDFTKKMLGRIFEVDLNILEDEELQASSFCLKHCKRLNKLRLSVSSHILERDLEILETSKFDSRMHAWNSICSTLVTNENLHELDLSNSKLHASSVKGLCLALKNPRCKVQKLTCKSVTPEWVLQDLIIALQGNSKLTHLNFSSNKLGMTVPLILKALRHSACNLKYLCLEKCNLSAASCQDLALFLTSIQHVTRLCLGFNRLQDDGIKLLCAALTHPKCALERLELWFCQLAAPACKHLSDALLQNRSLTHLNLSKNSLRDEGVKFLCEALGRPDGNLQSLNLSGCSFTREGCGELANALSHNHNVKILDLGENDLQDDGVKLLCEALKPHRALHTLGLAKCNLTTACCQHL | |
RUO | |
NLRP13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |