Learn More
Abnova™ Human NKRF Full-length ORF (AAH68514.1, 1 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055922-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm. [provided by RefSeq]
Sequence: MEKILQMAEGIDIGEMPSYDLVLSKPSKGQKRHLSTCDGQNPPKKQAGSKFHARPRFEPVHFVASSSKDERQEDPYGPQTKEVNEQTHFASMPRDIYQDYTQDSFSIQDGNSQYCDSSGFILTKDQPVTANMYFDSGNPAPSTTSQQANSQSTPEPSPSQTFPESVVAEKQYFIEKLTATIWKNLSNPEMTSGSDKINYTYMLTRCIQACKTNPEYIYAPLKEIPPADIPKNKKLLTDGYACEVRCQNIYLTTGYAGSKNGSRDRATELAVKLLQKRIEVRVVRRKFKHTFGEDLVVCQIGMSSYEFPPALKQPEDLVVLGKDASGQPIFNASAKHWTNFVITENANDAIGILNNSASFNKMSIEYKYEMMPNRTWRCRVFLQDHCLAEGYGTKKTSKHAAADEALKILQKTQPTYPSVKSSQCHTGSSPRGSGKKKDIKDLVVYENSSNPVCTLNDTAQFNRMTVEYVYERMTGLRWKCKVILESEVIAEAVGVKKTVKYEAAGEAVKTLKKTQPTVINNLKKGAVEDVISRNEIQGRSAEEAYKQQIKEDNIGNQLLRKMGWTGGGLGKSGEGIREPISVKEQHKREGLGLDVERVNKIAKRDIEQIIRNYARSESHTDLTFSRELTNDERKQIHQIAQKYGLKSKSHGVGHDRYLVVGRKRRKEDLLDQLKQEGQVGHYELVMPQANSpecifications
AAH68514.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
104.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ITBA4/NRF | |
NKRF | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55922 | |
NKRF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEKILQMAEGIDIGEMPSYDLVLSKPSKGQKRHLSTCDGQNPPKKQAGSKFHARPRFEPVHFVASSSKDERQEDPYGPQTKEVNEQTHFASMPRDIYQDYTQDSFSIQDGNSQYCDSSGFILTKDQPVTANMYFDSGNPAPSTTSQQANSQSTPEPSPSQTFPESVVAEKQYFIEKLTATIWKNLSNPEMTSGSDKINYTYMLTRCIQACKTNPEYIYAPLKEIPPADIPKNKKLLTDGYACEVRCQNIYLTTGYAGSKNGSRDRATELAVKLLQKRIEVRVVRRKFKHTFGEDLVVCQIGMSSYEFPPALKQPEDLVVLGKDASGQPIFNASAKHWTNFVITENANDAIGILNNSASFNKMSIEYKYEMMPNRTWRCRVFLQDHCLAEGYGTKKTSKHAAADEALKILQKTQPTYPSVKSSQCHTGSSPRGSGKKKDIKDLVVYENSSNPVCTLNDTAQFNRMTVEYVYERMTGLRWKCKVILESEVIAEAVGVKKTVKYEAAGEAVKTLKKTQPTVINNLKKGAVEDVISRNEIQGRSAEEAYKQQIKEDNIGNQLLRKMGWTGGGLGKSGEGIREPISVKEQHKREGLGLDVERVNKIAKRDIEQIIRNYARSESHTDLTFSRELTNDERKQIHQIAQKYGLKSKSHGVGHDRYLVVGRKRRKEDLLDQLKQEGQVGHYELVMPQAN | |
RUO | |
NKRF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |