missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NISCH Partial ORF (AAH56900, 1246 a.a. - 1345 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011188-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106342

  • 528.00€ / 25µg

Estimated Shipment: 03-11-2023
to see stock.
Add to basket

Description

Description

Sequence: LTGSTPMQVVTCLTRDSYLTHCFLQHLMVVLSSLERTPSPEPVDKDFYSEFGNKTTGKMENYELIHSSRVKFTYPSEEEIGDLTFTVAQKMAEPEKAPAL
Specifications

Specifications

AAH56900
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LTGSTPMQVVTCLTRDSYLTHCFLQHLMVVLSSLERTPSPEPVDKDFYSEFGNKTTGKMENYELIHSSRVKFTYPSEEEIGDLTFTVAQKMAEPEKAPAL
RUO
NISCH
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11188
NISCH (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14425/FLJ40413/FLJ90519/I-1/IRAS/KIAA0975
NISCH
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers