missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NID1 Partial ORF (NP_002499.1, 1148 a.a. - 1247 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004811-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121935

  • 508.00€ / 25µg
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix. [provided by RefSeq]

Sequence: EGLQYPFAVTSYGKNLYFTDWKMNSVVALDLAISKETDAFQPHKQTRLYGITTALSQCPQGHNYCSVNNGGCTHLCLATPGSRTCRCPDNTLGVDCIERK
Specifications

Specifications

NP_002499.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EGLQYPFAVTSYGKNLYFTDWKMNSVVALDLAISKETDAFQPHKQTRLYGITTALSQCPQGHNYCSVNNGGCTHLCLATPGSRTCRCPDNTLGVDCIERK
RUO
NID1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4811
NID1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NID
NID1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers