missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NHLH2 Partial ORF (NP_005590.1, 36 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004808-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: SDLEPVEEAEGDGKGGSRAALYPHPQQLSREEKRRRRRATAKYRSAHATRERIRVEAFNLAFAELRKLLPTLPPDKKLSKIEILRLAICYISYLNHVLDVSpecifications
NP_005590.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
SDLEPVEEAEGDGKGGSRAALYPHPQQLSREEKRRRRRATAKYRSAHATRERIRVEAFNLAFAELRKLLPTLPPDKKLSKIEILRLAICYISYLNHVLDV | |
RUO | |
NHLH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4808 | |
NHLH2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HEN2/KIAA0490/NSCL2/bHLHa34 | |
NHLH2 | |
Recombinant | |
wheat germ expression system |