missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NFYA Full-length ORF (NP_002496.1, 1 a.a. - 347 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004800-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100738

  • 349.00€ / 10µg

Estimated Shipment: 11-07-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms. [provided by RefSeq]

Sequence: MEQYTANSNSSTEQIVVQAGQIQQQQQGGVTAVQLQTEAQVASASGQQVQTLQVVQGQPLMVQVSGGQLITSTGQPIMVQAVPGGQGQTIMQVPVSGTQGLQQIQLVPPGQIQIQGGQAVQVQGQQGQTQQIIIQQPQTAVTAGQTQTQQQIAVQGQQVAQTAEGQTIVYQPVNADGTILQQVTVPVSGMITIPAASLAGAQIVQTGANTNTTSSGQGTVTVTLPVAGNVVNSGGMVMMVPGAGSVPAIQRIPLPGAEMLEEEPLYVNAKQYHRILKRRQARAKLEAEGKIPKERRKYLHESRHRHAMARKRGEGGRFFSPKEKDSPHMQDPNQADEEAMTQIIRVS
Specifications

Specifications

NP_002496.1
Liquid
4800
NFYA (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEQYTANSNSSTEQIVVQAGQIQQQQQGGVTAVQLQTEAQVASASGQQVQTLQVVQGQPLMVQVSGGQLITSTGQPIMVQAVPGGQGQTIMQVPVSGTQGLQQIQLVPPGQIQIQGGQAVQVQGQQGQTQQIIIQQPQTAVTAGQTQTQQQIAVQGQQVAQTAEGQTIVYQPVNADGTILQQVTVPVSGMITIPAASLAGAQIVQTGANTNTTSSGQGTVTVTLPVAGNVVNSGGMVMMVPGAGSVPAIQRIPLPGAEMLEEEPLYVNAKQYHRILKRRQARAKLEAEGKIPKERRKYLHESRHRHAMARKRGEGGRFFSPKEKDSPHMQDPNQADEEAMTQIIRVS
RUO
NFYA
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CBF-A/CBF-B/FLJ11236/HAP2/NF-YA
NFYA
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers