Learn More
Abnova™ Human NFKBIZ Full-length ORF (NP_113607.1, 1 a.a. - 718 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064332-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the ankyrin-repeat family and is induced by lipopolysaccharide (LPS). The C-terminal portion of the encoded product which contains the ankyrin repeats, shares high sequence similarity with the I kappa B family of proteins. The latter are known to play a role in inflammatory responses to LPS by their interaction with NF-B proteins through ankyrin-repeat domains. Studies in mouse indicate that this gene product is one of the nuclear I kappa B proteins and an activator of IL-6 production. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MIVDKLLDDSRGGEGLRDAAGGCGLMTSPLNLSYFYGASPPAAAPGACDASCSVLGPSAPGSPGSDSSDFSSASSVSSCGAVESRSRGGARAERQPVEPHMGVGRQQRGPFQGVRVKNSVKELLLHIRSHKQKASGQAVDDFKTQGVNIEQFRELKNTVSYSGKRKGPDSLSDGPACKRPALLHSQFLTPPQTPTPGESMEDVHLNEPKQESSADLLQNIINIKNECSPVSLNTVQVSWLNPVVVPQSSPAEQCQDFHGGQVFSPPQKCQPFQVRGSQQMIDQASLYQYSPQNQHVEQQPHYTHKPTLEYSPFPIPPQSPAYEPNLFDGPESQFCPNQSLVSLLGDQRESENIANPMQTSSSVQQQNDAHLHSFSMMPSSACEAMVGHEMASDSSNTSLPFSNMGNPMNTTQLGKSLFQWQVEQEESKLANISQDQFLSKDADGDTFLHIAVAQGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLLKNKSLVDTIKCLIQMGAAVEAKDRKSGRTALHLAAEEANLELIRLFLELPSCLSFVNAKAYNGNTALHVAASLQYRLTQLDAVRLLMRKGADPSTRNLENEQPVHLVPDGPVGEQIRRILKGKSIQQRAPPYSpecifications
NP_113607.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
104.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30225/FLJ34463/IKBZ/INAP/MAIL | |
NFKBIZ | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64332 | |
NFKBIZ (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIVDKLLDDSRGGEGLRDAAGGCGLMTSPLNLSYFYGASPPAAAPGACDASCSVLGPSAPGSPGSDSSDFSSASSVSSCGAVESRSRGGARAERQPVEPHMGVGRQQRGPFQGVRVKNSVKELLLHIRSHKQKASGQAVDDFKTQGVNIEQFRELKNTVSYSGKRKGPDSLSDGPACKRPALLHSQFLTPPQTPTPGESMEDVHLNEPKQESSADLLQNIINIKNECSPVSLNTVQVSWLNPVVVPQSSPAEQCQDFHGGQVFSPPQKCQPFQVRGSQQMIDQASLYQYSPQNQHVEQQPHYTHKPTLEYSPFPIPPQSPAYEPNLFDGPESQFCPNQSLVSLLGDQRESENIANPMQTSSSVQQQNDAHLHSFSMMPSSACEAMVGHEMASDSSNTSLPFSNMGNPMNTTQLGKSLFQWQVEQEESKLANISQDQFLSKDADGDTFLHIAVAQGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLLKNKSLVDTIKCLIQMGAAVEAKDRKSGRTALHLAAEEANLELIRLFLELPSCLSFVNAKAYNGNTALHVAASLQYRLTQLDAVRLLMRKGADPSTRNLENEQPVHLVPDGPVGEQIRRILKGKSIQQRAPPY | |
RUO | |
NFKBIZ | |
Wheat Germ (in vitro) | |
GST | |
Liquid |