Description
NFE2L2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. This cell line contains three unique indels: -101bp, -103bp, and -1bp deletion in exon 4 of the NFE2L2 gene. The same indel patterns are picked up in cDNA sequencing. Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 1,000,000 Cells |
| Regulatory Status | RUO |