Learn More
Abcam Human NFE2L2 knockout A549 cell line[21]
Product Code. 32157345 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32157345 1,000,000 Cells Each
32139435 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32157345 Supplier Abcam Supplier No. ab2853591000000Cells_
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
NFE2L2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. This cell line contains three unique indels: -101bp, -103bp, and -1bp deletion in exon 4 of the NFE2L2 gene. The same indel patterns are picked up in cDNA sequencing. Knockout. To ord...
NFE2L2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. This cell line contains three unique indels: -101bp, -103bp, and -1bp deletion in exon 4 of the NFE2L2 gene. The same indel patterns are picked up in cDNA sequencing. Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells