Learn More
Abnova™ Human NETO1 Partial ORF (NP_620416.1, 434 a.a. - 533 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00081832-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants encoding distinct proteins have been described. [provided by RefSeq]
Sequence: GSQLSSTKGSRSNLSTRDASILTEMPTQPGKPLIPPMNRRNILVMKHNYSQDAADACDIDEIEEVPTTSHRLSRHDKAVQRFCLIGSLSKHESEYNTTRVSpecifications
NP_620416.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GSQLSSTKGSRSNLSTRDASILTEMPTQPGKPLIPPMNRRNILVMKHNYSQDAADACDIDEIEEVPTTSHRLSRHDKAVQRFCLIGSLSKHESEYNTTRV | |
RUO | |
NETO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
81832 | |
NETO1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCTL1/BTCL1 | |
NETO1 | |
Recombinant | |
wheat germ expression system |