missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Nephrin DuoSet ELISA
Click to view available options
Sufficient For:
Five 96-well plates
Unit Size:
Pack of 5
Description
The Human Nephrin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Nephrin.
Specifications
Specifications
| Assay Range | 0.37 to 90pg/mL |
| Immunoassay Kit Format | 96-well Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | Five 96-well plates |
| Target Species | Human |
| Gene ID (Entrez) | 4868 (Human); 54631 (Mouse); 64563 (Rat) |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Sample Volume | 100 μL |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction